Learn More
Meso Scale Discovery V-PLEX Rat IL-5 Kit
SDP
Catalog No. 5000082429
Encompass
The V-PLEX Rat IL-5 Kit from Meso Scale Discovery is an immunoassay that uses electrochemiluminescence (ECL) for the quantitative measurement of interleukin 5 (IL-5) in rat serum plasma and cell culture supernatants The assay has high sensitivity wide dynamic range and requires low sample volume The kit includes a 96-well plate coated with capture antibody detection antibody conjugated to an ECL label calibrators diluent wash buffer read buffer and plate seals The assay takes approximately 3 5 hours to complete after a 2 hour incubation The kit has a detection range of 1 53-2500 pg/mL and includes reagents for 1 x 96-well plate
Catalog No. 50-000-82429 Supplier Meso Scale Discovery Supplier No. K153QSD4
Add to Cart
Add to Cart
Request a Quote
The V-PLEX Rat IL-5 Kit from Meso Scale Discovery is an immunoassay that uses electrochemiluminescence (ECL) for the quantitative measurement of interleukin 5 (IL-5) in rat serum plasma and cell culture supernatants The assay has high sensitivity wide dynamic range and requires low sample volume The kit includes a 96-well plate coated with capture antibody detection antibody conjugated to an ECL label calibrators diluent wash buffer read buffer and plate seals The assay takes approximately 3 5 hours to complete after a 2 hour incubation The kit has a detection range of 1 53-2500 pg/mL and includes reagents for 1 x 96-well plate