Learn More
Meso Scale Discovery V-PLEX Rat IL-5 Kit
SDP
Catalog No. 5000081561
Encompass
The Meso Scale Discovery V-PLEX Plus Rat IL-5 Kit is an electrochemiluminescent immunoassay for the quantitative measurement of IL-5 in rat serum plasma and cell culture supernatants The kit contains a 96-well plate coated with capture antibodies detection antibodies conjugated to an electrochemiluminescent label calibrators diluent wash buffer read buffer and plate seals The assay has a detection range of 1 53-2500 pg/mL and takes approximately 3 5 hours to run It uses an instrument such as the MSD QuickPlex SQ 120 for plate reading The kit includes sufficient reagents for 1 x 96-well plate when run as described
Catalog No. 50-000-81561 Supplier Meso Scale Discovery Supplier No. K153QSD1
The Meso Scale Discovery V-PLEX Plus Rat IL-5 Kit is an electrochemiluminescent immunoassay for the quantitative measurement of IL-5 in rat serum plasma and cell culture supernatants The kit contains a 96-well plate coated with capture antibodies detection antibodies conjugated to an electrochemiluminescent label calibrators diluent wash buffer read buffer and plate seals The assay has a detection range of 1 53-2500 pg/mL and takes approximately 3 5 hours to run It uses an instrument such as the MSD QuickPlex SQ 120 for plate reading The kit includes sufficient reagents for 1 x 96-well plate when run as described