Learn More
Meso Scale Discovery V-PLEX Rat IL-6 Kit, IL-6, Rat, Single-analyte, Sensitivity 13.8 pg/mL, Dynamic Range 13.8 - 8,550 pg/mL, 96-well plate
SDP
Catalog No. NC2235714
Encompass
V-PLEX Rat IL-6 Kit, IL-6, Rat, Single-analyte, Sensitivity 13.8 pg/mL, Dynamic Range 13.8 - 8,550 pg/mL, 96-well plate
This item is not returnable. View return policy
Catalog No. NC2235714 Supplier Meso Scale Discovery Supplier No. K153QXD2
V-PLEX Rat IL-6 Kit, IL-6, Rat, Single-analyte, Sensitivity 13.8 pg/mL, Dynamic Range 13.8 - 8,550 pg/mL, 96-well plate