Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Meso Scale Discovery V-PLEX Rat IL-6 Kit, Single-analyte immunoassay, 96-well plate, LLOD 13.8 pg/mL, Dynamic Range 13.8 - 8,550 pg/mL, Sample Types: Serum, Plasma, Urine, Cell Culture Supernatant
SDP

Catalog No. NC2284826
Encompass

The V-PLEX Rat IL-6 Kit is a high-performance immunoassay designed for quantitatively measuring rat Interleukin-6 (IL-6) in various biological samples. This kit utilizes the validated V-PLEX platform to ensure accurate results and includes a broad dynamic range and high sensitivity for precise quantification across multiple sample types.

  • Analyte: IL-6
  • Species: Rat
  • Assay type: Single-analyte immunoassay
  • Platform: V-PLEX
  • Plate type: 96-well
  • Sensitivity (LLOD): 13.8 pg/mL
  • Dynamic range: 13.8 - 8,550 pg/mL
  • Sample types: Serum, plasma, urine, cell culture supernatant
  • Capture antibody: Mouse monoclonal
  • Detection antibody: Goat polyclonal
  • Recombinant standards: Expressed in insect cell line
  • Usage: For research use only

This item is not returnable. View return policy
Catalog No. NC2284826 Supplier Meso Scale Discovery Supplier No. K153QXD4
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

The V-PLEX Rat IL-6 Kit is a high-performance immunoassay designed for quantitatively measuring rat Interleukin-6 (IL-6) in various biological samples. This kit utilizes the validated V-PLEX platform to ensure accurate results and includes a broad dynamic range and high sensitivity for precise quantification across multiple sample types.

  • Analyte: IL-6
  • Species: Rat
  • Assay type: Single-analyte immunoassay
  • Platform: V-PLEX
  • Plate type: 96-well
  • Sensitivity (LLOD): 13.8 pg/mL
  • Dynamic range: 13.8 - 8,550 pg/mL
  • Sample types: Serum, plasma, urine, cell culture supernatant
  • Capture antibody: Mouse monoclonal
  • Detection antibody: Goat polyclonal
  • Recombinant standards: Expressed in insect cell line
  • Usage: For research use only

This item is not returnable. View return policy
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.