Learn More
Meso Scale Discovery V-PLEX Rat KC/GRO Kit
SDP
Catalog No. 5000082297
Encompass
The V-PLEX Rat KC/GRO Kit from Meso Scale Discovery is a multiplex immunoassay for measuring rat keratinocyte chemoattractant (KC) and rat growth-related oncogene (GRO) in serum plasma and cell culture supernatants It includes a 96-well plate with capture antibodies detection antibodies Read Buffer Diluent 2 Calibrator Diluent and lyophilized standards for standard curves The electrochemiluminescent assay offers high sensitivity and a broad dynamic range enabling efficient quantification of KC and GRO from a small sample volume
Catalog No. 50-000-82297 Supplier Meso Scale Discovery Supplier No. K153QTD4
Add to Cart
Add to Cart
Request a Quote
The V-PLEX Rat KC/GRO Kit from Meso Scale Discovery is a multiplex immunoassay for measuring rat keratinocyte chemoattractant (KC) and rat growth-related oncogene (GRO) in serum plasma and cell culture supernatants It includes a 96-well plate with capture antibodies detection antibodies Read Buffer Diluent 2 Calibrator Diluent and lyophilized standards for standard curves The electrochemiluminescent assay offers high sensitivity and a broad dynamic range enabling efficient quantification of KC and GRO from a small sample volume