Learn More
Meso Scale Discovery V-PLEX Rat TNF-a Kit
SDP
Catalog No. 5000080310
Encompass
The V-PLEX Rat TNF-alpha Kit from Meso Scale Discovery is an electrochemiluminescent immunoassay for measuring TNF-alpha in rat serum plasma and cell culture supernatants It uses MSD s V-PLEX technology is highly sensitive with a dynamic range of 15 6-2000 pg/mL and shows no cross-reactivity with other cytokines The kit includes a 96-well plate detection antibody Read Buffer Calibrator Diluent and TNF-alpha calibrator The assay takes about 3 5 hours It provides a rapid robust and ultra-sensitive method for quantifying TNF-alpha in rat samples
Catalog No. 50-000-80310 Supplier Meso Scale Discovery Supplier No. K153QWD1
The V-PLEX Rat TNF-alpha Kit from Meso Scale Discovery is an electrochemiluminescent immunoassay for measuring TNF-alpha in rat serum plasma and cell culture supernatants It uses MSD s V-PLEX technology is highly sensitive with a dynamic range of 15 6-2000 pg/mL and shows no cross-reactivity with other cytokines The kit includes a 96-well plate detection antibody Read Buffer Calibrator Diluent and TNF-alpha calibrator The assay takes about 3 5 hours It provides a rapid robust and ultra-sensitive method for quantifying TNF-alpha in rat samples