Learn More
Meso Scale Discovery V-PLEX Viral Panel 1 NHP Kit
SDP
Catalog No. 5000081508
Encompass
The Meso Scale Discovery V-PLEX Viral Panel 1 NHP Kit is a multiplex immunoassay kit for the simultaneous detection of antibodies to SARS-CoV-2 Influenza A Influenza B and RSV in non-human primate serum plasma and lung lavage samples The kit contains a 96-well plate coated with antigens for the viruses detection antibodies assay diluent wash buffer read buffer and calibrators to quantify antibody levels The electrochemiluminescence immunoassay allows high sensitivity and a broad dynamic range The kit can help monitor immune responses to infection or vaccination in non-human primate studies
Catalog No. 50-000-81508 Supplier Meso Scale Discovery Supplier No. K15348D4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery V-PLEX Viral Panel 1 NHP Kit is a multiplex immunoassay kit for the simultaneous detection of antibodies to SARS-CoV-2 Influenza A Influenza B and RSV in non-human primate serum plasma and lung lavage samples The kit contains a 96-well plate coated with antigens for the viruses detection antibodies assay diluent wash buffer read buffer and calibrators to quantify antibody levels The electrochemiluminescence immunoassay allows high sensitivity and a broad dynamic range The kit can help monitor immune responses to infection or vaccination in non-human primate studies