Learn More
Meso Scale Discovery V-PLEX Viral Panel 1 NHP Kit
SDP
Catalog No. 5000081307
Encompass
The Meso Scale Discovery V-PLEX Viral Panel 1 (NHP) Kit is a multiplex immunoassay for detecting antibodies to SARS-CoV-2 Influenza A Influenza B and RSV in non-human primate serum and plasma It uses electrochemiluminescence on patterned arrays for high sensitivity and specificity The kit includes a 96-well plate with antigens detection antibodies wash buffers controls calibrators and reagents The assay takes about 3 hours and provides quantitative results for antibody levels enabling efficient viral antibody profiling from small samples
Catalog No. 50-000-81307 Supplier Meso Scale Discovery Supplier No. K15348D2
Add to Cart
Add to Cart
The Meso Scale Discovery V-PLEX Viral Panel 1 (NHP) Kit is a multiplex immunoassay for detecting antibodies to SARS-CoV-2 Influenza A Influenza B and RSV in non-human primate serum and plasma It uses electrochemiluminescence on patterned arrays for high sensitivity and specificity The kit includes a 96-well plate with antigens detection antibodies wash buffers controls calibrators and reagents The assay takes about 3 hours and provides quantitative results for antibody levels enabling efficient viral antibody profiling from small samples