Learn More
Meso Scale Discovery V-PLEX Viral Panel 2 Human Kit, Multiplex immunoassay, 5 each, Sub to low pg/mL detection limit
SDP
Catalog No. NC2256064
Encompass
The MESO SCALE DISCOVERY V-PLEX Viral Panel 2 Human Kit is an analytically validated immunoassay kit designed for the quantitative measurement of seven biomarkers associated with the human immune response to viral infection. Leveraging Electrochemiluminescence (ECL) technology, this kit provides accurate, reproducible results with high lot-to-lot consistency and detection limits in the sub- to low pg/mL range, making it suitable for rigorous research applications.
  • Comprehensive biomarker detection for seven key biomarkers.
  • Validated performance under rigorous design controls.
  • High reproducibility for consistent results across assay runs.
  • Exceptional sensitivity with low detection limits.
  • Flexible assay formats supporting both multiplex and singleplex configurations.
This item is not returnable. View return policy
Catalog No. NC2256064 Supplier Meso Scale Discovery Supplier No. K15346D2
Add to Cart
Add to Cart
Request a Quote
The MESO SCALE DISCOVERY V-PLEX Viral Panel 2 Human Kit is an analytically validated immunoassay kit designed for the quantitative measurement of seven biomarkers associated with the human immune response to viral infection. Leveraging Electrochemiluminescence (ECL) technology, this kit provides accurate, reproducible results with high lot-to-lot consistency and detection limits in the sub- to low pg/mL range, making it suitable for rigorous research applications.
  • Comprehensive biomarker detection for seven key biomarkers.
  • Validated performance under rigorous design controls.
  • High reproducibility for consistent results across assay runs.
  • Exceptional sensitivity with low detection limits.
  • Flexible assay formats supporting both multiplex and singleplex configurations.