Learn More
Meso Scale Discovery V-PLEX Viral Panel 2 NHP Kit
SDP
Catalog No. 5000079851
Encompass
The Meso Scale Discovery V-PLEX Viral Panel 2 (NHP) Kit is a multiplex immunoassay that simultaneously detects antibodies to SARS-CoV-2 Influenza A Influenza B and RSV in non-human primate serum and plasma samples This assay utilizes electrochemiluminescence detection technology on patterned arrays for high sensitivity and specificity The kit includes a 96-well plate coated with antigens detection antibodies wash buffers controls calibrators and other reagents needed to run the assay It can quantitatively measure antibody levels and has applications in vaccine development antiviral drug discovery and immunology research using non-human primate models
Catalog No. 50-000-79851 Supplier Meso Scale Discovery Supplier No. K15349D4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery V-PLEX Viral Panel 2 (NHP) Kit is a multiplex immunoassay that simultaneously detects antibodies to SARS-CoV-2 Influenza A Influenza B and RSV in non-human primate serum and plasma samples This assay utilizes electrochemiluminescence detection technology on patterned arrays for high sensitivity and specificity The kit includes a 96-well plate coated with antigens detection antibodies wash buffers controls calibrators and other reagents needed to run the assay It can quantitatively measure antibody levels and has applications in vaccine development antiviral drug discovery and immunology research using non-human primate models