Learn More
Meso Scale Discovery V-PLEX Viral Panel 2 NHP Kit
SDP
Catalog No. 5000080379
Encompass
The Meso Scale Discovery V-PLEX Viral Panel 2 (NHP) Kit is a multiplex immunoassay that simultaneously detects antibodies to SARS-CoV-2 Influenza A Influenza B and RSV in non-human primate serum and plasma samples This assay utilizes MSD s electrochemiluminescence detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements It includes a 96-well plate coated with antigens for the viruses an MSD Read Buffer and an MSD Discovery Workbench for analysis The kit enables quantification of antibodies that bind to the viral antigens
Catalog No. 50-000-80379 Supplier Meso Scale Discovery Supplier No. K15349D1
The Meso Scale Discovery V-PLEX Viral Panel 2 (NHP) Kit is a multiplex immunoassay that simultaneously detects antibodies to SARS-CoV-2 Influenza A Influenza B and RSV in non-human primate serum and plasma samples This assay utilizes MSD s electrochemiluminescence detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements It includes a 96-well plate coated with antigens for the viruses an MSD Read Buffer and an MSD Discovery Workbench for analysis The kit enables quantification of antibodies that bind to the viral antigens