Learn More
Meso Scale Discovery V-PLEX Viral Panel 3 NHP Kit
SDP
Catalog No. 5000079379
Encompass
The Meso Scale Discovery V-PLEX Viral Panel 3 NHP Kit is a multiplex immunoassay that simultaneously detects antibodies to SARS-CoV-2 Influenza A Virus and Influenza B Virus in non-human primate serum and plasma samples This assay utilizes electrochemiluminescence detection technology and provides quantitative results for each analyte in a single well The kit includes all necessary reagents and plates for analysis of up to 80 samples in duplicate This product enables efficient viral antibody profiling and is suitable for vaccine and therapeutic development studies involving non-human primates
Catalog No. 50-000-79379 Supplier Meso Scale Discovery Supplier No. K15350D2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery V-PLEX Viral Panel 3 NHP Kit is a multiplex immunoassay that simultaneously detects antibodies to SARS-CoV-2 Influenza A Virus and Influenza B Virus in non-human primate serum and plasma samples This assay utilizes electrochemiluminescence detection technology and provides quantitative results for each analyte in a single well The kit includes all necessary reagents and plates for analysis of up to 80 samples in duplicate This product enables efficient viral antibody profiling and is suitable for vaccine and therapeutic development studies involving non-human primates