Learn More
Meso Scale Discovery V-PLEX Viral Panel 3 NHP Kit
SDP
Catalog No. 5000081359
Encompass
The Meso Scale Discovery V-PLEX Viral Panel 3 NHP Kit is a multiplex immunoassay that simultaneously detects antibodies to SARS-CoV-2 Influenza A Virus and Influenza B Virus in non-human primate serum and plasma samples This assay utilizes electrochemiluminescence detection technology and provides quantitative results in under 5 hours The kit includes a 96-well plate coated with antigens for the viruses detection antibodies wash buffers controls and other necessary reagents for performing the assay This product enables efficient serological testing of non-human primates for current or prior exposure to these viruses
Catalog No. 50-000-81359 Supplier Meso Scale Discovery Supplier No. K15350D4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery V-PLEX Viral Panel 3 NHP Kit is a multiplex immunoassay that simultaneously detects antibodies to SARS-CoV-2 Influenza A Virus and Influenza B Virus in non-human primate serum and plasma samples This assay utilizes electrochemiluminescence detection technology and provides quantitative results in under 5 hours The kit includes a 96-well plate coated with antigens for the viruses detection antibodies wash buffers controls and other necessary reagents for performing the assay This product enables efficient serological testing of non-human primates for current or prior exposure to these viruses