Learn More
Meso Scale Discovery V-PLEX Viral Panel 3 NHP Kit
SDP
Catalog No. 5000082047
Encompass
The Meso Scale Discovery V-PLEX Viral Panel 3 NHP Kit is a multiplex immunoassay kit that detects antibodies to SARS-CoV-2 Influenza A and Influenza B in non-human primate serum and plasma samples The kit contains a 96-well plate coated with antigens for the viruses detection antibodies buffers calibrators and controls The electrochemiluminescence assay allows for high sensitivity and a broad dynamic range Results are obtained in under 5 hours using an MSD instrument This kit enables efficient serological testing of non-human primates
Catalog No. 50-000-82047 Supplier Meso Scale Discovery Supplier No. K15350D1
The Meso Scale Discovery V-PLEX Viral Panel 3 NHP Kit is a multiplex immunoassay kit that detects antibodies to SARS-CoV-2 Influenza A and Influenza B in non-human primate serum and plasma samples The kit contains a 96-well plate coated with antigens for the viruses detection antibodies buffers calibrators and controls The electrochemiluminescence assay allows for high sensitivity and a broad dynamic range Results are obtained in under 5 hours using an MSD instrument This kit enables efficient serological testing of non-human primates