Learn More
Meso Scale Discovery V-PLEX Plus Mouse Cytokine 19-Plex Kit, 19 analytes measured, multiplex assay format, detection range sub- to low pg/mL, includes controls, wash buffer, plate seals.
SDP
Catalog No. NC2890351
Encompass
V-PLEX Plus Mouse Cytokine 19-Plex Kit, 19 analytes measured, multiplex assay format, detection range sub- to low pg/mL, includes controls, wash buffer, plate seals.
Catalog No. NC2890351 Supplier Meso Scale Discovery Supplier No. K15255G1
V-PLEX Plus Mouse Cytokine 19-Plex Kit, 19 analytes measured, multiplex assay format, detection range sub- to low pg/mL, includes controls, wash buffer, plate seals.