Learn More
Meso Scale Discovery V-PLEX Plus Proinflammatory Panel1 Mouse Kit, 5 plates
SDP
Catalog No. NC1161297
Encompass
The Proinflammatory Panel 1 (mouse) Kit provides assay-specific components for the quantitative determination of IFN-gamma, IL-1beta, IL-2, IL-4, IL-5, IL-6, KC/GRO, IL-10, IL-12p70, and TNF-alpha. This kit may be used for the analysis of all or any combination of these cytokine biomarkers in cell culture supernatants, serum, plasma, and urine.
Catalog No. NC1161297 Supplier Meso Scale Discovery Supplier No. K15048G2
Add to Cart
Add to Cart
Request a Quote
The Proinflammatory Panel 1 (mouse) Kit provides assay-specific components for the quantitative determination of IFN-gamma, IL-1beta, IL-2, IL-4, IL-5, IL-6, KC/GRO, IL-10, IL-12p70, and TNF-alpha. This kit may be used for the analysis of all or any combination of these cytokine biomarkers in cell culture supernatants, serum, plasma, and urine.