Learn More
Analytical Sales & Services V-Trough for Multi-Channel Pipetors, 12 Channel
Catalog No. 5000806198
Encompass
Advantage Clear Polystyrene 12 Channel V-Trough for Multi-Channel Pipetors 3mL Volume per Channel
Qty/Pkg: 5
Catalog No. 50-008-06198 Supplier Analytical Sales & Services Supplier No. 96013
Advantage Clear Polystyrene 12 Channel V-Trough for Multi-Channel Pipetors 3mL Volume per Channel
Qty/Pkg: 5