Learn More
American Radiolabeled Chemicals Inc V1A ANTA 125I 50UC
SDP
Catalog No. 50901027
Encompass
Vasopressin (linear), V1A antagonist (Phenylacetyl1-0-Me-D-Tyr2) Arg6-125I 50uCi in buffered solution (pH 4.2), 0.05 mCi/ml, S.A. 2200 Ci/mmol, M.W. 1274
Catalog No. 50-901-027 Supplier American Radiolabeled Chemicals Inc Supplier No. ARI 014550UC
Add to Cart
Add to Cart
Request a Quote
Vasopressin (linear), V1A antagonist (Phenylacetyl1-0-Me-D-Tyr2) Arg6-125I 50uCi in buffered solution (pH 4.2), 0.05 mCi/ml, S.A. 2200 Ci/mmol, M.W. 1274