Learn More
Eurofins Discoverx Ready-to-Assay V 1A Vasopressin Receptor Frozen Cells, 2 Vials of Cells. (HTS059RTA)
Catalog No. 502005773
Encompass
The Ready-to-Assay V 1A Vasopressin Receptor Frozen Cells are a simple, thaw-and-assay cell format for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This assay assesses ligand (e.g. vasopressin) based activation of AVPR1A GPCR activity via detection of increasing levels of intracellular calcium. The included cell line overexpresses AVPR1A alongside endogenous high levels of a promiscuous G-protein, coupling the receptor to the calcium signaling pathway. Upon activation of AVPR1A, the resulting calcium activity can be fluorescently assayed. Ready-to-assay kits consist of two vials of the stably modified clonal cell line in a convenient thaw-and-use format. For detailed product information, User manual and datasheet, visit www.discoverx.com/products-applications/frozen-cells.
Catalog No. 50-200-5773 Supplier Eurofins Discoverx Supplier No. HTS059RTA
The Ready-to-Assay V 1A Vasopressin Receptor Frozen Cells are a simple, thaw-and-assay cell format for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This assay assesses ligand (e.g. vasopressin) based activation of AVPR1A GPCR activity via detection of increasing levels of intracellular calcium. The included cell line overexpresses AVPR1A alongside endogenous high levels of a promiscuous G-protein, coupling the receptor to the calcium signaling pathway. Upon activation of AVPR1A, the resulting calcium activity can be fluorescently assayed. Ready-to-assay kits consist of two vials of the stably modified clonal cell line in a convenient thaw-and-use format. For detailed product information, User manual and datasheet, visit www.discoverx.com/products-applications/frozen-cells.