Learn More
Lexogen V2 LIBRARY PREP KIT FWD WITH U
SDP
Catalog No. NC2479730
Encompass
  • Library Preparation Kit for 3' mRNA-Seq
  • Streamlines library preparation workflow in less than 4.5 hours
  • No prior poly(A) enrichment required
  • No rRNA depletion necessary
  • Suitable for various sample types, including low-input and degraded RNA (down to 1 ng)
  • Incorporates patented 12 nt Unique Dual Indices (UDI)
  • Allows multiplexing of up to 96 samples
  • Compatible with Illumina NGS platforms (iSeq, MiniSeq, NextSeq, HiSeq, NovaSeq)
  • Includes Library Generation, Amplification, and Purification Modules
  • Strand specificity confirmed
  • RNA fragmentation not required
  • Improved PCR system for library quality and resistance to overcycling
Catalog No. NC2479730 Supplier Lexogen Supplier No. 191.96
  • Library Preparation Kit for 3' mRNA-Seq
  • Streamlines library preparation workflow in less than 4.5 hours
  • No prior poly(A) enrichment required
  • No rRNA depletion necessary
  • Suitable for various sample types, including low-input and degraded RNA (down to 1 ng)
  • Incorporates patented 12 nt Unique Dual Indices (UDI)
  • Allows multiplexing of up to 96 samples
  • Compatible with Illumina NGS platforms (iSeq, MiniSeq, NextSeq, HiSeq, NovaSeq)
  • Includes Library Generation, Amplification, and Purification Modules
  • Strand specificity confirmed
  • RNA fragmentation not required
  • Improved PCR system for library quality and resistance to overcycling