Learn More
Perkin Elmer US LLC V21 Wear Metal Standard, 900 µg/g, in Hydrocarbon Oil, 100 g
Catalog No. 502104008
Encompass_Preferred
High quality reference materials are essential for accurate analytical measurement and quality control, ensuring sound decisions are made based on reliable data. PerkinElmer offers a full range of wear metal standards for all of your application needs.
Catalog No. 50-210-4008 Supplier Perkin Elmer US LLC Supplier No. N9308312
High quality reference materials are essential for accurate analytical measurement and quality control, ensuring sound decisions are made based on reliable data. PerkinElmer offers a full range of wear metal standards for all of your application needs.