Learn More
Pyramex™ V2G-Plus™ Safety Eyewear
Catalog No. 19155347 Shop All Pyramex Safety Products Products
Change view
Click to view available options
Lens Tint:
Clear
Gray
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Lens Tint
19-155-347 Clear
19-155-348 Gray
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-155-347 Supplier Pyramex™ Supplier No. GB6410SDT
Add to Cart
Add to Cart
This item is not returnable. View return policy
H2X anti-fog dual lenses
  • Includes a wide adjustable elastic strap with quick release system for secure fit
  • TPU frame features excellent resistance to cold and heat and provides perfect facial seal
  • Special indirect ventilation system located at the top and bottom of frame allow air flow and heat release
  • Multi-functional goggle with 180º vision, protects agaisnt chemical splash, sun glare, dust, flying particles, wind, and 99.9% of harmful UV rays
  • Outer polycarbonate lens protects against the environment, while an inner lens prevents fogging
  • Inner lens is made from acetate, separated by a thin spacer of air, sealed together with foam
  • Thermo Lens System provides excellent resistance to fogging caused by temperature difference between indoor and outdoor or between on-site and ambient environment
  • Passes MIL-PRF 32432 High Velocity Impact Standards
  • Rx Insert available (RX6400)
  • Specifications
    Lens Coating Anti-fog
    Lens Type H2X
    Certifications/Compliance ANSI Z87.1,CSA Z94.3-07
    Certifications/Compliance (Europe) EN-166
    Lens Tint Clear
    Product Type Goggles
    Frame Color Black
    Style Goggle with Strap
    Weight (Metric) 80 g
    Lens Material Polycarbonate (Front Lens), Acetate (Inner Lens), PU Foam
    Frame Material TPU