Learn More
Pyramex™ V2G™ Readers Safety Eyewear
Catalog No. 19155332 Shop All Pyramex Safety Products Products
Change view
Click to view available options
Magnification Power:
+1.50
+2.00
+2.50
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Magnification Power
19-155-332 +2.50
19-155-330 +1.50
19-155-331 +2.00
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 19-155-332 Supplier Pyramex™ Supplier No. GB1810STR25
Add to Cart
Add to Cart
This item is not returnable. View return policy
Available in three magnifying strengths
  • Flame resistant foam padding
  • Lens and frame vented to increase air flow
  • Interchangeable temples and headband included
  • 9.75 base curved lens provides full side protection
  • Headband and temples feature ratchet adjustment for perfect fit
  • Features quick fit retainer
  • Scratch resistant polycarbonate lens provides 99% UVA/B/C protection
  • Ballistic options pass MIL-PRF 32432 High Velocity Impact Standards
  • Specifications
    Lens Type H2X
    Certifications/Compliance ANSI Z87.1 ,CSA Z94.3-07
    Lens Tint Clear
    Frame Color Black
    Includes Strap
    Magnification Power +2.50
    Lens Material Polycarbonate
    Frame Material PVC