Learn More
Pyramex™ V2G™ Safety Eyewear
Catalog No. 19155337 Shop All Pyramex Safety Products Products
Change view
Click to view available options
Lens Tint:
Amber
Clear
Gray
IR/UV 3.0
IR/UV 5.0
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Lens Tint
19-155-337 IR/UV 3.0
19-155-329 Clear
19-155-334 Gray
19-155-335 Amber
19-155-336 IR/UV 5.0
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 19-155-337 Supplier Pyramex™ Supplier No. GB1860SFT
Add to Cart
Add to Cart
This item is not returnable. View return policy
Black frame with temples and strap
  • Flame resistant foam padding
  • Lens and frame vented to increase air flow
  • Interchangeable temples and headband included
  • 9.75 base curved lens provides full side protection
  • Headband and temples feature ratchet adjustment for perfect fit
  • Features quick fit retainer
  • Scratch resistant polycarbonate lens provides 99% UVA/B/C protection
  • Ballistic options pass MIL-PRF 32432 High Velocity Impact Standards
  • Specifications
    Lens Type H2X
    Certifications/Compliance ANSI Z87.1
    Description Ballistic
    Lens Tint IR/UV 3.0
    Frame Color Black
    Weight (Metric) 38 g
    Lens Material Polycarbonate
    Frame Material PVC