Learn More
Pyramex™ V3T™ Safety Eyewear
Catalog No. 19155644 Shop All Pyramex Safety Products Products
Change view
Click to view available options
Lens Tint:
Amber
Clear
Gray
Indoor/Outdoor, Mirror
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Lens Tint
19-155-644 Indoor/Outdoor, Mirror
19-155-641 Clear
19-155-642 Gray
19-155-643 Amber
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 19-155-644 Supplier Pyramex™ Supplier No. SB10380ST
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
This item is not returnable. View return policy
Anti-fog lenses
  • Vented lens and frame
  • Quick release strap
  • Flame resistant foam padding
  • Passes MIL-PRF 32432 High Velocity Impact Standards
  • Specifications
    Certifications/Compliance ANSI Z87.1,CSA Z94.3
    Lens Tint Indoor/Outdoor, Mirror
    Frame Color Black
    Weight (Metric) 51.72 g
    Lens Material Polycarbonate
    Frame Material Nylon