Learn More
Cytek Biosciences Inc V420 Anti-Human CD3, 100 tests, cFluor™ V420 fluorophore, Clone SK7, Reactivity Human, Chimpanzee, Flow Cytometry
Supplier: Cytek Biosciences Inc R720053
The Cytek Biosciences cFluor™ V420 Anti-Human CD3 (SK7) is a high-quality monoclonal antibody intended for research use only. Designed for detailed immunophenotyping of human and chimpanzee samples with flow cytometry, this antibody targets the epsilon chain of the human CD3 antigen and is conjugated with the cFluor™ V420 fluorophore for robust performance. Each product lot undergoes rigorous quality control testing, ensuring reliable results in applications requiring precise antibody properties.
- Monoclonal antibody for human and chimpanzee reactivity
- Targets human CD3 epsilon chain
- Conjugated with cFluor™ V420 fluorophore
- Purified via affinity chromatography
- 100 tests per vial
- Application: Flow cytometry
- Test volume: 5 μL / test
- Storage: 2-8°C, protected from light
- Do not freeze
- Antigen molecular weight: 20 to 30 kDa
- Quality control via flow cytometric analysis
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.