Learn More
Cytek Biosciences Inc vF 450 aMu CD127 Anti Primary
Supplier: Cytek Biosciences Inc 751271U100
Product Name: violetFluor 450 Anti-Mouse CD127 (IL-7Ra) (A7R34)
Product Class: Antibody
Type: Primary
Product Name: violetFluor 450 Anti-Mouse CD127 (IL-7Ra) (A7R34)
Cytek Catalog Number: 75-1271-U100
Marker: CD127 (IL-7Ra)
Target Species: Mouse
Clonality: Monoclonal
Clone: A7R34
Host Species: Rat
Isotype: IgG2a, k
Conjugation: violetFluor 450
Size: 100 μg
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.