Learn More
Apexbio Technology LLC V5 Epitope Tag Peptide, 285983-48-4, MFCD09752957, 5 mg
SDP
Supplier:  Apexbio Technology LLC A60055
Encompass_Preferred
MF: C64H108N16O20, MW: 1421.64, Solubility: 71.082mg/mL in DMSO, 107.2 mg/mL in EtOH, 55.4 mg/mL in H2O. The recognized V5 epitope represents 95GKPIPNPLLGLDST108 of RNA polymerase alpha subunit of simian parainuenza virus type 5. This short peptide sequence was chosen because high-afnity antibodies can be reliably produced in many different species.
Catalog No. 50-115-0869