Learn More
Medchemexpress LLC HY-P0325 1mg Medchemexpress, V5 Epitope Tag Peptide Trifluoroacetate CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP03251MG
Encompass_Preferred
Medchemexpress, HY-P0325 1mg V5 Epitope Tag Peptide Trifluoroacetate CAS: V5 Epitope Tag Peptide Trifluoroacetate is a tag peptide derived from a small epitope present on the P and V proteins of the paramyxovirus of simian virus 5. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4120