Learn More
Enzo Life Sciences V5-tag Recombinant monoclonal antibody (SV5-P-K) (Human IgG1 kappa) (200µg)
SDP
Supplier:  Enzo Life Sciences ENZABS5910200
Encompass_Preferred
Clone: SV5-P-K. Isotype: IgG1κ. Immunogen: Pk (V5) Epitope Tag (GKPIPNPLLGLDST). Species reactivity: Virus. Specificity: Recognizes a 14 aa sequence (GKPIPNPLLGLDST) derived from the P and V proteins of the Paramyxovirus simian virus 5. Applications: ELISA, IF, IHC (FS), IP, WB Radioimmunoassay. Purity: Protein A affinity purified. Formulation: Liquid. In PBS containing 0.02% Proclin 300.
Catalog No. 50-201-3117