Learn More
Sino Biological V5 Synthetic Peptide 1 mg
Supplier:  Sino Biological PP1003781
Encompass_Preferred
V5 peptide can be used to competitively elute V5-tagged fusion proteins from immobilized anti-V5 magnetic beads. Use V5 peptide to elute V5-tagged proteins. Elution yields will increase by incubating the peptide on the tube for 10-15 minutes at 30-37 oC for each elution fraction.
Catalog No. 50-006-96274