Learn More
Sino Biological V5 tag PAb 20ul (Sino Biological)
Supplier:  Sino Biological 100378T3620
Encompass_Preferred
Produced in rabbits immunized with purified, a synthetic peptide corresponding to the V5-tag sequence. V5-tag specific IgG was purified by V5-taq affinity chromatography.
Catalog No. 50-237-2240