Learn More
Proteintech Group Inc V5-Trap Agarose
SDP
Catalog No. 502258894 Shop All Proteintech Group Inc Products
Encompass
V5-Trap Agarose is an affinity reagent for IP and purification of V5-tagged proteins. It consists of an anti-V5 Nanobody/ VHH coupled to agarose beads.
This item is not returnable. View return policy
Catalog No. 50-225-8894 Supplier Proteintech Group Inc Supplier No. V5TA10
V5-Trap Agarose is an affinity reagent for IP and purification of V5-tagged proteins. It consists of an anti-V5 Nanobody/ VHH coupled to agarose beads.