Learn More
Proteintech Group Inc V5-Trap Magnetic Agarose
SDP
Catalog No. 502258909 Shop All Proteintech Group Inc Products
Encompass
The ChromoTek V5-Trap Magnetic Agarose is an affinity resin for IP of V5-tagged proteins. It consists of a anti V5 Nanobody/VHH coupled to magnetic agarose beads.
This item is not returnable. View return policy
Catalog No. 50-225-8909 Supplier Proteintech Group Inc Supplier No. V5TMA20
The ChromoTek V5-Trap Magnetic Agarose is an affinity resin for IP of V5-tagged proteins. It consists of a anti V5 Nanobody/VHH coupled to magnetic agarose beads.