Learn More
Proteintech Group Inc V5-Trap Magnetic Particles M-270
SDP
Catalog No. 502258901 Shop All Proteintech Group Inc Products
Encompass
The ChromoTek V5-Trap Magnetic Particles M-270 consists of an anti-V5-tag Nanobody (VHH), which is covalently bound to Magnetic Particles M-270. V5-Trap Magnetic Particles M-270 is used to immunoprecipitate V5-tagged fusion proteins from cell extracts of various organisms like mammals, plants, bacteria, yeast, insects etc.
This item is not returnable. View return policy
Catalog No. 50-225-8901 Supplier Proteintech Group Inc Supplier No. V5TD10
The ChromoTek V5-Trap Magnetic Particles M-270 consists of an anti-V5-tag Nanobody (VHH), which is covalently bound to Magnetic Particles M-270. V5-Trap Magnetic Particles M-270 is used to immunoprecipitate V5-tagged fusion proteins from cell extracts of various organisms like mammals, plants, bacteria, yeast, insects etc.