Learn More
Cytek Biosciences Inc vF 500 aMu CD45.2 Anti Primary
Supplier: Cytek Biosciences Inc 850454U025
Product Name: violetFluor 500 Anti-Mouse CD45.2 (104)
Product Class: Antibody
Type: Primary
Product Name: violetFluor 500 Anti-Mouse CD45.2 (104)
Cytek Catalog Number: 85-0454-U025
Marker: CD45.2
Target Species: Mouse
Clonality: Monoclonal
Clone: 104
Host Species: Mouse
Isotype: IgG2a, k
Conjugation: violetFluor 500
Size: 25 μg
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.