Learn More
MOLTOX Molecular Toxicology Inc V8 Agar, 20 mL/plates, 20 plates/sleeve, Store in Refrigerator
SDP
Catalog No. 50948669
Encompass_Preferred
V8 Agar is used in the cultivation and sporation of fungi. Each lot of media from MOLTOX is tested for sterility, where applicable, and its ability to support the growth of the appropriate microorganism. All media are accompanied by GLP-compliant formulation and Quality Control certificate.
Catalog No. 50-948-669 Supplier MOLTOX Molecular Toxicology Inc Supplier No. 2140S49
V8 Agar is used in the cultivation and sporation of fungi. Each lot of media from MOLTOX is tested for sterility, where applicable, and its ability to support the growth of the appropriate microorganism. All media are accompanied by GLP-compliant formulation and Quality Control certificate.