Learn More
Gilson Company Inc 8" Sieve, All Stainless, Full Height, No. 635
Catalog No. NC9504014
Encompass_Preferred
8in (203mm) diameter ASTM E11 Test Sieve has stainless steel woven-wire cloth with No. 635 (20µm) openings and full height stainless steel frame. The overall height is 2-5/8in and the stacked height is 2-1/8in. This sieve is optionally available fitted with #35 (500µm) stainless steel backing cloth for enhanced durability.
Catalog No. NC9504014 Supplier Gilson Company Inc Supplier No. V8SF #635
8in (203mm) diameter ASTM E11 Test Sieve has stainless steel woven-wire cloth with No. 635 (20µm) openings and full height stainless steel frame. The overall height is 2-5/8in and the stacked height is 2-1/8in. This sieve is optionally available fitted with #35 (500µm) stainless steel backing cloth for enhanced durability.