Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hardy Diagnostics™ V9 Agar for mycology
SDP

Catalog No. 23001816 Shop All Hardy Diagnostics Products
Change view
Click to view available options
Product Type: Agar Plate
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
23-001-816 Product Type: Agar Plate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-001-816 Supplier Hardy Diagnostics™ Supplier No. G98
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

(Potato Flake Agar with V8 Juice) V9 Agar is recommended for the isolation, cultivation and rapid sporulation of fungi

  • Optically clear polystyrene for distortion free viewing.
  • The naturally low pH makes the media inhibitory to most bacteria

Specifications

Appearance Opaque and Granular
Certifications/Compliance ISO 13485
Color Orange
Content And Storage 2°C to 8°C
Detects Fungi, Yeasts, Molds
Form Prepared Medium
Format Plate
Media Type V9 Agar
Product Type Agar Plate
Sterility Sterile
Target Organism Class Fungi
Test Type Fungal Testing
Fill Volume 22 mL
Formulation Dehydrated Potato Flakes - 20.0 gm, V8 Juice - 350.0 mL, Agar - 20.0 gm per 650 mL deionized water
For Use With (Application) Fungi Cultivation, Sporulation, Isolation
Incubation Conditions 30°C
Incubation Time 7 days
Material Polystyrene
No. per Pack 10
Packaging Type Breathable Bag
Quantity 10 Ea.
Dimensions (Dia. x H) 100 x 15 mm
Height (Metric) 15 mm
Diameter (Metric) 100 mm
Species Trichophyton rubrum
Source V9 Agar (V8 juice and potato flakes)
Manufacturing Quality ISO 13485
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.