Learn More
Teledyne Tekmar V9 TRAP
Catalog No. NC9793961
Encompass
The Stratum PTC ships with a #9 proprietary U-shaped trap installed and a U-shaped Vocarb 3000 trap. If a sample is not properly desorbed from the trap, the resolution in the chromatogram will suffer. The U-shaped trap provides superior peak shape by allowing a volume for the desorbing gas to refocus before proceeding to the GC. The result is a dramatic improvement in your chromatographic resolution.
Catalog No. NC9793961 Supplier Teledyne Tekmar Supplier No. 149908403
The Stratum PTC ships with a #9 proprietary U-shaped trap installed and a U-shaped Vocarb 3000 trap. If a sample is not properly desorbed from the trap, the resolution in the chromatogram will suffer. The U-shaped trap provides superior peak shape by allowing a volume for the desorbing gas to refocus before proceeding to the GC. The result is a dramatic improvement in your chromatographic resolution.