Learn More
PDC VA Direct Thermal Paper Labels, 2500/Roll, 4 Rolls/Box
Catalog No. 21235819
Change view
Click to view available options
Quantity:
2500/Roll, 4 Rolls/Box
Catalog No. Quantity
21-235-819 2500/Roll, 4 Rolls/Box
1 options
Catalog No. 21-235-819 Supplier PDC Supplier No. TD112XP
  • $212.00
    $158.65 / Case of 4
    Save $53.35
    25% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
White labels with strong adhesive, designed for reliable, high-volume labeling in clinical, laboratory, and hospital settings.
  • Engineered for dependable performance in demanding clinical environments
  • Extra permanent adhesive ensures labels stay securely in place on various surfaces
  • White face stock provides clear, legible printing without the need for ribbons
  • Compatible with most direct thermal printers for efficient workflow
  • Ideal for high-volume applications in hospitals, laboratories, and healthcare facilities
Specifications
Adhesive Type Permanent
Coating None
Color White
Finish Matte
Format Label
Material Paper
Printable Sides Front
Print Technology Direct Thermal
Product Type Label
Sterility Non-sterile
Quantity 2500/Roll, 4 Rolls/Box
Dimensions (L x W) 2 x 1 in.
Length (English) 2 in.
Width (English) 1 in.
Width (Metric) 25.4 mm