Learn More
Metrohm USA 663 VA Stand for Metrohm Autolab potentiostats
SDP
Supplier:  Metrohm USA 26630020
Encompass
The 663 VA Stand is a voltammetric measuring stand for potentiostats from Metrohm Autolab. The 663 VA Stand can be used to perform polarographic and voltammetric measurements on the following electrodes: Multi-Mode electrode pro (MME), rotating disk electrodes or scTRACE Gold.
Catalog No. NC1808501
Add to Cart
Request a Quote