Learn More
Enzo Life Sciences Vα TCR Recombinant monoclonal antibody (34) (Mouse IgG1 kappa) (200µg)
SDP
Catalog No. 502013132
Encompass_Preferred
Clone: 34. Isotype: IgG1κ. Immunogen: Purified recombinant soluble 85.33 scTCR. Species reactivity: Human. Applications: ELISA, Flow Cytometry, WB. Purity: Protein A affinity purified. Formulation: Liquid. In PBS containing 0.02% Proclin 300.
Catalog No. 50-201-3132 Supplier Enzo Life Sciences Supplier No. ENZABS6060200
Add to Cart
Add to Cart
Clone: 34. Isotype: IgG1κ. Immunogen: Purified recombinant soluble 85.33 scTCR. Species reactivity: Human. Applications: ELISA, Flow Cytometry, WB. Purity: Protein A affinity purified. Formulation: Liquid. In PBS containing 0.02% Proclin 300.