Learn More
Instech Vascular Access Button™ for Mouse, 2 Channel Oval Base, 14mm silicone mesh, 25ga/25ga, with Injector
SDP
Catalog No. NC2816923
Encompass
The VABM2JBSM/25R25 is a two-channel Vascular Access Button™ designed for mice, featuring a 14mm silicone mesh disk to promote tissue growth for long-term stability. Developed with The Jackson Laboratory, this VAB™ allows for simultaneous infusion and blood sampling or glucose clamping. The closed system uses PinPorts™ to enable aseptic manual access while preventing catheter exposure, which improves patency and reduces evaporation. Mice can be group-housed post-surgery when used with a protective aluminum cap.
Features
  • Material: 14mm silicone mesh disk for tissue integration
  • Connectors: Two 25ga catheter connectors facing the sides
  • Application: Glucose clamping and simultaneous sampling
  • Compatibility: VABM2T/25 tethers and 25ga swivels
  • Housing: Supports group housing with a protective cap
  • Access: Manual access via PinPorts™ using PNP3M injectors
Catalog No. NC2816923 Supplier Instech Supplier No. VABM2JBSM/25R25
The VABM2JBSM/25R25 is a two-channel Vascular Access Button™ designed for mice, featuring a 14mm silicone mesh disk to promote tissue growth for long-term stability. Developed with The Jackson Laboratory, this VAB™ allows for simultaneous infusion and blood sampling or glucose clamping. The closed system uses PinPorts™ to enable aseptic manual access while preventing catheter exposure, which improves patency and reduces evaporation. Mice can be group-housed post-surgery when used with a protective aluminum cap.
Features
  • Material: 14mm silicone mesh disk for tissue integration
  • Connectors: Two 25ga catheter connectors facing the sides
  • Application: Glucose clamping and simultaneous sampling
  • Compatibility: VABM2T/25 tethers and 25ga swivels
  • Housing: Supports group housing with a protective cap
  • Access: Manual access via PinPorts™ using PNP3M injectors