Learn More
Instech Vascular Access Button™ for Mouse, 2 Channel 25ga/25ga, Oval Base with Felt, with Injector
SDP
Supplier:  Instech VABM2JB/25R25
Encompass
Two channel magnetic Vascular Access Button™ for mice, 14mm polyester surgical felt disk, oval base, 25ga red port and 25ga white port catheter connectors (for 2Fr or 1-2Fr catheters) facing tail. Injector included.
The mouse VAB™ is a transcutaneous button that permits quick, aseptic access to a catheter.
Catalog No. NC2816922