Learn More
Instech Tether for Magnetic 2 Channel Rat Vascular Access Button, 12in (30cm)
SDP
Catalog No. 501004895
Encompass
Two channel rat Vascular Access Button tether assembly, 12in, sterile. Mates with VAR2B/22 button. For continuous infusion and blood sampling or bile sampling.
Catalog No. 50-100-4895 Supplier Instech Supplier No. VABR2T/25
Two channel rat Vascular Access Button tether assembly, 12in, sterile. Mates with VAR2B/22 button. For continuous infusion and blood sampling or bile sampling.