Learn More
Instech Mating tether for Vascular Access Button™. Polyurethane tubing. 12in (30cm) long. Mates with 375/22PS or stainless steel swivels. For rat infusion and blood sampling. Sterile
SDP
Catalog No. 5081239
Encompass
The Instech VAB95T is the tether assembly designed for use with VAB95‑series rat Vascular Access Buttons, providing a secure, aseptic connection for continuous infusion or blood sampling. It mates with the button’s external port by twisting into place, forming a closed, low‑dead‑volume fluid path that minimizes contamination and supports reliable long‑term catheter access during tethered studies.
Features
  • Tether assembly for VAB95‑series rat access buttons
  • Twist‑in connection for secure continuous access
  • Closed fluid path to reduce contamination risk
  • Supports continuous infusion and sampling workflows
  • Compatible with standard 3Fr polyurethane catheter systems
Catalog No. 50-812-39 Supplier Instech Supplier No. VAB95T
The Instech VAB95T is the tether assembly designed for use with VAB95‑series rat Vascular Access Buttons, providing a secure, aseptic connection for continuous infusion or blood sampling. It mates with the button’s external port by twisting into place, forming a closed, low‑dead‑volume fluid path that minimizes contamination and supports reliable long‑term catheter access during tethered studies.
Features
  • Tether assembly for VAB95‑series rat access buttons
  • Twist‑in connection for secure continuous access
  • Closed fluid path to reduce contamination risk
  • Supports continuous infusion and sampling workflows
  • Compatible with standard 3Fr polyurethane catheter systems