Learn More
Medchemexpress LLC Vabicaserin hydrochloride | 887258-94-8 | MFCD31706226 | 99.5% | 264.79 | 100 MG
SDP
Catalog No. 5000343100
Encompass_Preferred
Vabicaserin hydrochloride is a 5-hydroxytryptamine 2C (5-HT2C) receptor-selective agonist with an EC50 of 8 nM. It acts as a potent agonist, stimulating 5-HT2C receptor-coupled calcium mobilization, and also exhibits 5-HT2A receptor antagonism and 5-HT2B antagonist or partial agonist activity.
  • Highly selective for 5-HT2C receptor.
  • Potent agonist for 5-HT2C receptor-coupled calcium mobilization.
  • Exhibits 5-HT2A receptor antagonism and 5-HT2B antagonist/partial agonist activity.
  • Suitable for research use only.
  • High purity: 99.47%.
  • Documentation available, including Data Sheet, COA, SDS.
  • Detailed solubility protocols provided for in vitro and in vivo studies.
  • Supported by multiple scientific publications.
  • Associated with various compound libraries.
  • Investigated in clinical trials (Phase 1 and 2).
Catalog No. 50-003-43100 Supplier Medchemexpress LLC Supplier No. HY11120010MM/1ML
Vabicaserin hydrochloride is a 5-hydroxytryptamine 2C (5-HT2C) receptor-selective agonist with an EC50 of 8 nM. It acts as a potent agonist, stimulating 5-HT2C receptor-coupled calcium mobilization, and also exhibits 5-HT2A receptor antagonism and 5-HT2B antagonist or partial agonist activity.
  • Highly selective for 5-HT2C receptor.
  • Potent agonist for 5-HT2C receptor-coupled calcium mobilization.
  • Exhibits 5-HT2A receptor antagonism and 5-HT2B antagonist/partial agonist activity.
  • Suitable for research use only.
  • High purity: 99.47%.
  • Documentation available, including Data Sheet, COA, SDS.
  • Detailed solubility protocols provided for in vitro and in vivo studies.
  • Supported by multiple scientific publications.
  • Associated with various compound libraries.
  • Investigated in clinical trials (Phase 1 and 2).