Learn More
Medchemexpress LLC HY-19930 100mg , Vaborbactam CAS:1360457-46-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY19930100MG
Encompass_Preferred
HY-19930 100mg Vaborbactam CAS: 1360457-46-0 Vaborbactam (RPX7009) is a cyclic boronic acid pharmacophore β-lactamase inhibitor. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9387