Learn More
Medchemexpress LLC HY-19930 10mg Medchemexpress, Vaborbactam CAS:1360457-46-0 Purity:>98%
SDP
Catalog No. 502029302
Encompass_Preferred
Medchemexpress, HY-19930 10mg Vaborbactam CAS:1360457-46-0 Vaborbactam (RPX7009) is a cyclic boronic acid pharmacophore β-lactamase inhibitor. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9302 Supplier Medchemexpress LLC Supplier No. HY1993010MG
Medchemexpress, HY-19930 10mg Vaborbactam CAS:1360457-46-0 Vaborbactam (RPX7009) is a cyclic boronic acid pharmacophore β-lactamase inhibitor. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.